Water PCR tested for molecular biology DNA-free , suited for qPCR

Supplier: PanReac AppliChem

Synonyms: Hydrogen oxide

A8510.1017
APLIA8510.1017EA 969 SGD
APLIA8510.1017
Water PCR tested for molecular biology DNA-free , suited for qPCR
Water
Formula: H₂O
MW: 18.02 g/mol
Boiling Pt: 100
Melting Pt: 0
Density: 1 g/cm³ (4 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011332
CAS Number: 7732-18-5
EINECS: 231-791-2
Merck Index: 13,10098

Order Now

Specification Test Results

Bacterial DNA not detectable (min. 40 PCR cycles)
DNases/RNases not detectable


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR