SYBR® Green I staining reagent DNA-free

Supplier: PanReac AppliChem

Synonyms: Thiazole Green

A8511.50625
APLIA8511.50625EA 969 SGD
APLIA8511.50625
SYBR® Green I staining reagent DNA-free
SYBR® Green I

SYBR Green is an cyanine dye that acts for the detection of double stranded DNA. This ten fold concentrated dye solution is suitable for qPCR when primers such as 16S rDNA or 18S rDNA primers are used.


  • Concentration: 10X concentrated solution
  • Used for DNA gel electrophoresis
  • Lower mutagenic potential than ethidium bromide
  • Absorption maxima in the UV range are 284 nm and 382 nm


Mutagenicity can be increased by synergistic effects. The complex of DNA and SYBR Green absorbs blue light of wavelength 494 nm (absorption maximum) and emits green light at 521 nm (emission maximum).The stained DNA can be detected on a blue light transilluminator.


Avoid repeated freeze-thaw cycles! We recommend storing the product at 2-8°C after its first use.


SYBR is a registered trademark of Thermo Fisher Scientific.

Formula: C₃₂H₃₇N₄S
Storage Temperature: Freezer
MDL Number: MFCD00284715
CAS Number: 163795-75-3

Order Now

Specification Test Results

Bacterial DNA not detectable (min. 40 PCR cycles)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR