qPCR Mycoplasma Test Kit
Supplier: PanReac AppliChem
A9019.0025
APLIA9019.0025EA 2820 SGD
APLIA9019.0025
qPCR Mycoplasma Test Kit
Microbiology Test Systems Microorganism Tests
Lyophilised PCR mix for the detection of mycoplasma in cell culture by Real-Time/qPCR.
Delivery information: Supplied with Taq DNA polymerase. Kit contains PCR primer mix (incl. primers, probes, nucleotides), reaction buffer solution, water (PCR grade), positive template control, and internal control DNA.
Order Now
SPECIFICATIONS
Environmentally Preferable