Potassium permanganate 0.1 mol/l (0.5N) volumetric solution

Supplier: PanReac AppliChem
Danger

Synonyms: Permanganic acid potassium salt

182651.1611
APPC182651.1611EA 200 SGD
APPC182651.1611
Potassium permanganate 0.1 mol/l (0.5N) volumetric solution
Potassium permanganate

Use for the titration of percarbonates.

Formula: KMnO₄
MW: 158.03 g/mol
MDL Number: MFCD00011364
CAS Number: 7722-64-7
EINECS: 231-760-3
UN: 3214
ADR: 5.1 II
Merck Index: 12,07824

Order Now

Specification Test Results

Titer at 20 °C 0.999 - 1.001
Traceability NIST


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR