Picric acid, yellow, fluid

Supplier: PanReac AppliChem

Synonyms: 2,4,6-Trinitrophenol, TNP

251049.1610
APPC251049.1610EA 250 SGD
APPC251049.1610
Picric acid, yellow, fluid
Picric acid
Formula: C₆H₃N₃O₇
MW: 229.11
Boiling Pt: 100 °C (212 °F)
Melting Pt: 80.35
Density: 1,006 kg/L
Flash Pt: 150
Storage Temperature: Room Temperature
MDL Number: MFCD00007102
CAS Number: 88-89-1
Merck Index: 12,07562

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR