Automation-compatible PCR sealing mats, Axygen®
Supplier: Corning
AM-384-PCR-RD
AXYGAM-384-PCR-RDEA 93 SGD
AXYGAM-384-PCR-RD
Automation-compatible PCR sealing mats, Axygen®
Seals Microplate Seals
Axygen AxyMat sealing mats can be used in a wide range of temperatures for PCR and storage applications.
  • Minimize evaporation
  • Made of research-grade silicone
  • Autoclavable
They can be reused following a bleach wash and ethanol rinse protocol.
Order Now
SPECIFICATIONS
Environmentally Preferable