Semi-automated plate sealer, Axygen® PlateMax™
Supplier: Corning
HS-1120
AXYGHS-1120EA 12560 SGD
AXYGHS-1120
Semi-automated plate sealer, Axygen® PlateMax™
Heat Sealers Microplate Heat Sealers
PlateMax™ Semi Automated Plate Sealer is ideal heat sealer for the low to medium throughput Laboratory that requires uniform and consistent sealing of microplates
The PlateMax accepts a wide range of microplates and heat seals utilizing a control system with a large digital display that shows all operating parameters. The sealing temperatures are adjustable from 100°C to 190 °C and sealing times are adjustable from 0,5 to 10 seconds. The power saver feature that saves power by shutting down heating block when inactive, can now be toggled off for labs that wish to keep unit at sealingMore Product Information