Semi automated microplate heat sealer, Axygen® PlateMax®
Supplier: Corning
HS-1230
AXYGHS-1230EA 13350 SGD
AXYGHS-1230
Semi automated microplate heat sealer, Axygen® PlateMax®
Heat Sealers Microplate Heat Sealers
Axygen® PlateMax® semi automatic plate sealer is ideal for the low-to-medium throughput laboratory that requires uniform and consistent sealing of microplates. Offering complete versatility, the PlateMax® sealer will accept a full range of plates for PCR, assay or storage applications. It can be used to seal a wide range of plate heights. Sealing parameters are set and displayed via the user-friendly control panel and the sealing operation is automated to ensure consistent results.
  • Adjustable sealing temperature: 100 to 190 °C
  • Sealing times adjustable from
More Product Information