Mag plasmid kit, Axygen® AxyPrep™

Supplier: Corning

MAG-P-10
AXYGMAG-P-10EA 529 SGD
AXYGMAG-P-10
Mag plasmid kit, Axygen® AxyPrep™
Nucleic Acid Reagents Nucleic Acid Purification Kits and Reagents

The AxyPrep™ Mag Plasmid purification kit utilises a magnetic bead technology for high-throughput purification of plasmid DNA from E.coli cells.


AxyPrep™ Mag Plasmid kit can also be used with fosmid and BAC vector-based constructs ranging from 5 to 150kb.

Order Now

SPECIFICATIONS

Environmentally Preferable
Application
Target molecule Plasmid
Refine target molecule Plasmid
Sample type Bacteria
Method / Format Magnetic beads
Prep size Scalable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR