384 well clear V-bottom 240 µl, polypropylene, Axygen®
Supplier: Corning
P-384-240SQ-CS
AXYGP-384-240SQ-CSEA 604 SGD
AXYGP-384-240SQ-CS
384 well clear V-bottom 240 µl, polypropylene, Axygen®
Microplates Deep Well Microplates
Axygen® deep well and assay plates are used for sample collection, in vitro growth chambers and long-term storage. Made of polypropylene, they offer excellent chemical resistance and will withstand temperatures down to −80°C.
  • Excellent chemical resistance
  • ANSI/SLAS 1-2004: Microplates - Footprint Dimensions
  • Centrifugation to 4,000 RCF
  • Diamond shaped wells allow complete sample recovery
  • Black and white plates are available by special order
  • Ultra-flat surface ensures proper sealing with heat sealing film