Deep well plates, 24-well, Axygen®
Supplier: Corning
P-DW10ML24CS
AXYGP-DW10ML24CSEA 609 SGD
AXYGP-DW10ML24CS
Deep well plates, 24-well, Axygen®
Microplates Deep Well Microplates
Deep well and assay plates are used for sample collection, in vitro growth chambers and longterm storage. Made of PP, they offer excellent chemical resistance and will withstand temperatures down to –80 °C. Ultra-flat surface ensures proper sealing with heat sealing film. Clear V-shaped well bottom.