PCR tubes and caps, Axygen®
Supplier: Corning
PCR-02-A PCR-02-C PCR-02-L-C PCR-05-A PCR-05-C PCR-05-L-C PCR-02-C-500 PCR-05-C-500
AXYGPCR-02-AEA 95.7 SGD
AXYGPCR-02-A AXYGPCR-02-C AXYGPCR-02-L-C AXYGPCR-05-A AXYGPCR-05-C AXYGPCR-05-L-C AXYGPCR-02-C-500 AXYGPCR-05-C-500
PCR tubes and caps, Axygen®
Tubes PCR Tubes
Axygen thin wall PCR strip tubes are designed to fit a variety of thermal cyclers. The consistent and ultra-thin wall allows precise thermal transfer.
An even insertion cap ensures a tight fit to reduce evaporation. Strip tubes are available in an assortment of colors.
Optical flat cap strips feature ultra-clear upper surfaces, ideal for fluorescent luminescent and real-time PCR applications.
They offer a light transmittance that is 75% higher than with standard strip caps. Made to seal tube strips and 96-well microplates.
Order Now
SPECIFICATIONS
Environmentally Preferable Low Manufacturing Impact
Environmental certification I-RECs, ISO 14001 - Reynosa