PCR plates, 96-well, no skirt, Axygen®
Supplier: Corning
PCR-96-C PCR-96-C-S PCR-96-FLT-C
AXYGPCR-96-CEA 68.5 SGD
AXYGPCR-96-C AXYGPCR-96-C-S AXYGPCR-96-FLT-C
PCR plates, 96-well, no skirt, Axygen®
Microplates PCR Plates
PP plates, silicone mats. Designed to fit 0,2 ml thermal cycler blocks.
  • Ultra-thin walls of uniform thickness accelerate heat transfer, leading to shorter cycle times and more consistent results
  • Certified free from DNase, RNase and endotoxins
  • AxyMat™ sealing mats autoclavable and reusable
Volume with AxyMat™: 250 μl
Compatible with (in addition to standard thermal cycling):
Plates without skirt: Strategene MX4000, BioRad iCycler, MyiQ, iQ5
Plates without skirt, with flat top: ABI and BioRad qPCR systems
Plates without skirt, with elevated wells: Amersham MegaBACE sequencer
Order Now
SPECIFICATIONS
Environmentally Preferable Low Manufacturing Impact
Environmental certification I-RECs, ISO 14001 - Reynosa