PCR plates for Roche® LightCycler® 480, Axygen®
Supplier: Corning
PCR-96-LC480-W
AXYGPCR-96-LC480-WEA 927 SGD
AXYGPCR-96-LC480-W
PCR plates for Roche® LightCycler® 480, Axygen®
Microplates PCR Plates
PP, white. Low profile microplates compatible with many thermal cyclers, but especially suitable for the Roche® LightCycler® 480.