PCR plates, 96-well, segmented, Axygen®
Supplier: Corning
PCR-96-SG-C
AXYGPCR-96-SG-CEA 68.2 SGD
AXYGPCR-96-SG-C
PCR plates, 96-well, segmented, Axygen®
Microplates PCR Plates
PP plates, silicone mats. Designed to fit 0,2 ml thermal cycler blocks.
  • Ultra-thin walls of uniform thickness accelerate heat transfer, leading to shorter cycle times and more consistent results
  • Certified free from DNase, RNase and endotoxin for highest sample integrity
  • AxyMat™ sealing mats autoclavable and reusable
Volume with AxyMat™: 240 μl
Compatible with (in addition to standard thermal cycling): BioRad iCycler, MyiQ, iQ5, Beckman CEQ sequencers.