Microcentrifuge tubes, 0,5/1,5/2,0 ml, with screw caps, Axygen®
Supplier: Corning
SCT-200-SS-O SCT-200-SS-G SCT-050-A-S SCT-050-C-S SCT-050-SS-A-S SCT-050-SS-G SCT-050-SS-R SCT-150-A SCT-150-A-S SCT-150-C-S SCT-150-G SCT-150-SS-C-S SCT-200-A-S SCT-200-C-S SCT-200-SS-C-S ST-150-C-S ST-200-C-S SCT-050-SS-C SCT-150-C SCT-150-SS-C SCT-200-C SCT-200-SS-C SCT-200-SS-A-S SCT-050-SS-B-S SCT-050-SS-C-S SCT-050-SS-G-S SCT-050-SS-R-S SCT-050-SS-W-S SCT-050-SS-Y-S SCT-150-SS-A-S SCT-050-SS-O-S SCT-150-LP-C SCT-45-C-S ST-050-SS-S ST-200-SS-S SCT-050-B-S SCT-050-G-S SCT-050-R-S
AXYGSCT-200-SS-OEA 151 SGD
AXYGSCT-200-SS-O AXYGSCT-200-SS-G AXYGSCT-050-A-S AXYGSCT-050-C-S AXYGSCT-050-SS-A-S AXYGSCT-050-SS-G AXYGSCT-050-SS-R AXYGSCT-150-A AXYGSCT-150-A-S AXYGSCT-150-C-S AXYGSCT-150-G AXYGSCT-150-SS-C-S AXYGSCT-200-A-S AXYGSCT-200-C-S AXYGSCT-200-SS-C-S AXYGST-150-C-S AXYGST-200-C-S AXYGSCT-050-SS-C AXYGSCT-150-C AXYGSCT-150-SS-C AXYGSCT-200-C AXYGSCT-200-SS-C AXYGSCT-200-SS-A-S AXYGSCT-050-SS-B-S AXYGSCT-050-SS-C-S AXYGSCT-050-SS-G-S AXYGSCT-050-SS-R-S AXYGSCT-050-SS-W-S AXYGSCT-050-SS-Y-S AXYGSCT-150-SS-A-S AXYGSCT-050-SS-O-S AXYGSCT-150-LP-C AXYGSCT-45-C-S AXYGST-050-SS-S AXYGST-200-SS-S AXYGSCT-050-B-S AXYGSCT-050-G-S AXYGSCT-050-R-S
Microcentrifuge tubes, 0,5/1,5/2,0 ml, with screw caps, Axygen®
Tubes Microcentrifuge Tubes
PP, sterile or non sterile, conical base with or without skirt.
  • Easily perforated screw cap with extra long rim and O-ring to ensure effective, uniform closure for centrifugation
  • The rims of the tubes and caps are ribbed for easier opening and closing with one hand
  • Certified DNase-, RNase- and endotoxin-free
Designed for many laboratory applications such as the collection, treatment, centrifugation, long-term storage and transport of inhibitor enzymes or other reagents.
Volume: 0,5, 1,5 and 2,0 ml.
Max. RCF: 20000×g.
Delivery information: NonMore Product Information