Microcentrifuge tubes, Axygen®
Supplier: Corning
ST-150-SS ST-200-SS ST-050 ST-050-SS ST-050-SS-X ST-050-X ST-150 ST-150-SS-X ST-200 ST-200-SS-X ST-200-X ST-45 ST-45-S
AXYGST-150-SSEA 45.2 SGD
AXYGST-150-SS AXYGST-200-SS AXYGST-050 AXYGST-050-SS AXYGST-050-SS-X AXYGST-050-X AXYGST-150 AXYGST-150-SS-X AXYGST-200 AXYGST-200-SS-X AXYGST-200-X AXYGST-45 AXYGST-45-S
Microcentrifuge tubes, Axygen®
Tubes Microcentrifuge Tubes
Screw cap tubes made of PP feature a cap that provides a tight secure seal.
  • Certified RNase-/DNase-free, nonpyrogenic
  • Will withstand up to 20000×g
  • Screw caps to be ordered seperately
Screw cap tubes are available in a variety of colors with a conical or self-standing bottom.
Order Now
SPECIFICATIONS
Environmentally Preferable