Filter tips, Classic / Maxymum Recovery™, Axygen®
Supplier: Corning
TF-100 TF-1000 TF-1000-L-R-S TF-1000-R-S TF-1005-WB-R-S TF-100-R-S TF-150-R-S TF-20 TF-200 TF-200-R-S TF-205WB-RS TF-20-L-R-S TF-20-R-S TF-300 TF-300-L-R-S TF-300-R-S TF-350 TF-350-L-R-S TF-350-R-S TF-400-L-R-S TF-400-R-S TF-420-L-R-S TF-420-R-S TF-50 TF-50-L-R-S TF-50-R-S TXLF-10 TXLF-10-R-S TF-200-L-R-S TF-100-L-R-S TF-1200-C-R-S TF-400 TXLF-10-L-R-S
AXYGTF-100EA 114 SGD
AXYGTF-100 AXYGTF-1000 AXYGTF-1000-L-R-S AXYGTF-1000-R-S AXYGTF-1005-WB-R-S AXYGTF-100-R-S AXYGTF-150-R-S AXYGTF-20 AXYGTF-200 AXYGTF-200-R-S AXYGTF-205WB-RS AXYGTF-20-L-R-S AXYGTF-20-R-S AXYGTF-300 AXYGTF-300-L-R-S AXYGTF-300-R-S AXYGTF-350 AXYGTF-350-L-R-S AXYGTF-350-R-S AXYGTF-400-L-R-S AXYGTF-400-R-S AXYGTF-420-L-R-S AXYGTF-420-R-S AXYGTF-50 AXYGTF-50-L-R-S AXYGTF-50-R-S AXYGTXLF-10 AXYGTXLF-10-R-S AXYGTF-200-L-R-S AXYGTF-100-L-R-S AXYGTF-1200-C-R-S AXYGTF-400 AXYGTXLF-10-L-R-S
Filter tips, Classic / Maxymum Recovery™, Axygen®
Pipette Tips
PP, sterile or non sterile.
  • Free of DNA, RNase, DNase, pyrogens and metals
  • Maxymum Recovery™ tips have an ultra-smooth surface designed to reduce sample retention and sample loss, and to avoid sample denaturation
  • Sterilised by gamma radiation
Order Now
SPECIFICATIONS
Environmentally Preferable