PCR tubes, 0,5 ml, without cap, Axygen®
Supplier: Corning
TUB-310-NC
AXYGTUB-310-NCEA 150 SGD
AXYGTUB-310-NC
PCR tubes, 0,5 ml, without cap, Axygen®
Tubes PCR Tubes
PCR tube, made of PP, designed to fit standart 96 well blocks. Ultra-thin and consistent wall thickness allows precise thermal transfer and even insertion cap ensures a tight fit to prevent evaporation.
  • Certified RNase-/DNase-free
  • Nonpyrogenic
  • Nonsterile
  • Compatible with ABI-310 Instrument
Order Now
SPECIFICATIONS
Environmentally Preferable Low Manufacturing Impact
Environmental certification I-RECs, ISO 14001 - Reynosa