Sealing film for PCR plates, Axygen® UltraClear
Supplier: Corning
UC-500
AXYGUC-500EA 653 SGD
AXYGUC-500
Sealing film for PCR plates, Axygen® UltraClear
Seals Microplate Seals
For sealing 96 and 384-well PCR plates.
  • Film clarity allows for optical analysis during Real-Time PCR
  • Unique pressure sensitive adhesive that leaves no tacky residue
  • Functional temperature range −40 to +104 °C
Order Now
SPECIFICATIONS
Environmentally Preferable