Ampoule breakers
Supplier: SciLabware
AWY700 AWY702
AZLOAWY700EA 932 SGD
AZLOAWY700 AZLOAWY702
Ampoule breakers
Ampoule Openers
Ampoule breakers enables safe and easy breaking of glass ampoules.
  • Broken ampoule top is retained for safe disposal
  • Choice of size to suit all common glass ampoules