Specimen containers, with captive hinged cap, Azlon®
Supplier: SciLabware
BGG308
AZLOBGG308EA 22.3 SGD
AZLOBGG308
Specimen containers, with captive hinged cap, Azlon®
Sample Containers General Purpose Containers
LDPE, translucent.
  • Good chemical resistance
  • Lightweight and unbreakable in normal use
  • Can withstand temperatures up to 80 °C
Containers with captive hinged cap suitable for many applications.
Caution: *The product(s) on this page are outside the scope of the EU Directive 2019/904