Aspirator bottles, heavy-duty, with screw caps and spigot, Azlon®
Supplier: SciLabware
BNP10A
AZLOBNP10AEA 172 SGD
AZLOBNP10A
Aspirator bottles, heavy-duty, with screw caps and spigot, Azlon®
Bottles Aspirators
PP, octagonal with white screw cap and heavy-duty spigot.
  • Very good chemical resistance
  • Autoclavable at +121 °C*
  • Easy to fill and pour with the offset narrow mouth
Unique shape allows easy location and efficient use of bench space. Equipped with an integral carrying handle and moulded graduations for easy and quick volume assessment. Caps have moulded drill guides for easy location of probes and feed lines. Leakproof heavy-duty spigot opens and closes in a quarter turn.
* Closures and spigots should be removed before autoclaving.