Laboratory trays, sterilising, Azlon®
Supplier: SciLabware
TWR204
AZLOTWR204EA 81.6 SGD
AZLOTWR204
Laboratory trays, sterilising, Azlon®
Trays/Dishes/Pots Sterilising Trays/Pans
PP, white.
  • Robust and deep to accommodate a wide range of instruments etc.
  • Autoclavable
  • Accessory lids are white polystyrene and are not autoclavable