Ammonium carbonate, BAKER ANALYZED® ACS, J.T.Baker®

Supplier: Avantor
Warning

Synonyms: Salt of hartshorn

BAKR0015.1000EA 76.8 SGD
BAKR0015.1000
Ammonium carbonate, BAKER ANALYZED® ACS, J.T.Baker®
Ammonium carbonate
Formula: (NH₄)₂CO₃
MW: 96,09 g/mol
Melting Pt: 58 °C
Density: 1,5 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010890
CAS Number: 506-87-6
EINECS: 208-058-0
Merck Index: 13,00508

Order Now

Specification Test Results

Meets ACS Specifications
Assay (NH3) min. 30.0 %
Trace Impurities (in ppm)Chloride (Cl) max. 5
Trace Impurities (in ppm)Iron (Fe) max. 5
Trace Impurities (in ppm)Heavy Metals (as Pb) max. 5
Insoluble Matter max. 0.005 %
Non-volatile Matter max. 0.01 %
Sulfur Compounds (as SO4) max. 0.002 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR