Ammonium chloride, BAKER ANALYZED® ACS, J.T.Baker®

Supplier: Avantor
Warning

Synonyms: Ammonium muriate, Salmiac

BAKR0038.0500EA 65 SGD
BAKR0038.0500
Ammonium chloride, BAKER ANALYZED® ACS, J.T.Baker®
Ammonium chloride
Formula: NH₄Cl
Melting Pt: 335 °C
Density: 1,5256 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011420
CAS Number: 12125-02-9
EINECS: 235-186-4
Merck Index: 13,00510
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6e45c4909ae60875d5b9dca2f28f54f5c8028e64&vid=389479ce9f26d046837e12f5c47dd4bc7d47ec2b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Meets ACS Specifications. Meets Reagent Specificatons for testing USP/NF monographs.
Assay (argentometric titrn.) min. 99.5 %
Calcium (Ca) max. 0.001 %
Insoluble Matter max. 0.005 %
pH of 5% Solution at 25°C 4.5 - 5.5
Residue after Ignition max. 0.01 %
Sulfate (SO4) max. 0.002 %
Trace Impurities (in ppm)Iron (Fe) max. 2
Trace Impurities (in ppm)Magnesium (Mg) max. 5
Trace Impurities (in ppm)Heavy Metals (as Pb) max. 5
Trace Impurities (in ppm)Phosphate (PO4) max. 2


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR