Barium chloride dihydrate 99.0-101.0%, BAKER ANALYZED® ACS, J.T.Baker®

Supplier: Avantor
Danger

BAKR0045.1000EA 186 SGD
BAKR0045.1000
Barium chloride dihydrate 99.0-101.0%, BAKER ANALYZED® ACS, J.T.Baker®
Barium chloride dihydrate
Formula: BaCl₂·2H₂O
Boiling Pt: 1560 °C (1013 hPa)
Melting Pt: 960 °C
Density: 3,86 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149154
CAS Number: 10326-27-9
EINECS: 233-788-1
UN: 1564
ADR: 6.1,III
Merck Index: 14,00971

Order Now

Specification Test Results

Exceeds ACS Specifications
Assay 99.0 - 101.0 %
Calcium (Ca) max. 0.05 %
Iron (Fe) max. 2 ppm
Potassium (K) max. 0.0025 %
Sodium (Na) max. 0.005 %
Strontium (Sr) max. 0.1 %
Heavy Metals (as Pb) max. 5 ppm
Insoluble Matter max. 0.005 %
Loss on Drying at 150°C 14.0 - 16.0 %
Oxidizing Substances (as NO3) max. 0.005 %
pH of 5% Solution at 25°C 5.2 - 8.2


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR