Orthoboric acid, BAKER ANALYZED® ACS, J.T.Baker®

Supplier: Avantor
Danger

BAKR0055.1000EA 54.1 SGD
BAKR0055.1000
Orthoboric acid, BAKER ANALYZED® ACS, J.T.Baker®
ortho-Boric acid
Formula: B(OH)₃
Boiling Pt: 185 °C (1013 hPa)
Melting Pt: 160 °C
Density: 1,44 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011337
CAS Number: 10043-35-3
EINECS: 233-139-2
REACH: 01-2119486683-25
Merck Index: 13,01326
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=098dd700154daaf6338ebb270902bb2bc704f27d&vid=6eec7d529090dd4c4b966ed8cd8d057153c5a47b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Exceeds ACS Specifications
Assay min. 99.5 %
Arsenic (As) max. 1 ppm
Calcium (Ca) max. 0.005 %
Chloride (Cl) max. 0.001 %
Iron (Fe) max. 0.001 %
Heavy Metals (as Pb) max. 0.001 %
Insoluble in Methanol max. 0.005 %
Nonvolatile with Methanol max. 0.05 %
Phosphate (PO4) max. 0.001 %
Sulfate (SO4) max. 0.005 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR