Citric acid kit, anhydrous 99.5-100.5% (by anhydrous basis), powder USP, J.T.Baker®

Supplier: Avantor
Warning

Synonyms: 2-Hydroxypropane-1,2,3-tricarboxylic acid, Hydroxytricarballylic acid

BAKR0122-K1EA 4620 SGD
BAKR0122-K1
Citric acid kit, anhydrous 99.5-100.5% (by anhydrous basis), powder USP, J.T.Baker®
Citric acid

Certifications: All the products are cGMP produced and meet USP grade standards.

Formula: HOC(COOH)(CH₂COOH)₂
MW: 192.13 g/mol
Boiling Pt: 310 °C (1013 hPa)
Melting Pt: 149…151 °C
Density: 1.44 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00011669
CAS Number: 77-92-9
EINECS: 201-069-1
REACH: 01-2119457026-42-0000
Merck Index: 13,02350

Order Now

Specification Test Results

Meets U.S.P Requirements
GMP Manufactured Product
Kit Components Meet Requirements Passes Test
Bulk Pharmaceutical Chemical
This kit contains one each of Citric Acid, Anhydrous product code 0122-B1 and 0122-SA


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR