Lithium chloride 99, BAKER ANALYZED® ACS, J.T.Baker®

Supplier: Avantor
Warning

BAKR0156.1000EA 255 SGD
BAKR0156.1000
Lithium chloride 99, BAKER ANALYZED® ACS, J.T.Baker®
Lithium chloride
Formula: LiCl
MW: 42.39 g/mol
Boiling Pt: 1382
Melting Pt: 614
Density: 2.07 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011078
CAS Number: 7447-41-8
EINECS: 231-212-3
Merck Index: 13,05550

Order Now

Specification Test Results

Meets ACS Specifications
Assay min. 99 %
Barium (Ba) max. 0.003 %
Calcium (Ca) max. 0.01 %
Iron (Fe) max. 0.001 %
Potassium (K) max. 0.01 %
Sodium (Na) max. 0.20 %
Heavy Metals (as Pb) max. 0.002 %
Insoluble Matter max. 0.01 %
Loss on Drying max. 1.0 %
Sulfate (SO4) max. 0.01 %
Titrable Base (meq/g) max. 0.008

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR