Magnesium chloride hexahydrate, BAKER ANALYZED® ACS, J.T.Baker®

Supplier: Avantor

BAKR0162.1000EA 73 SGD
BAKR0162.1000
Magnesium chloride hexahydrate, BAKER ANALYZED® ACS, J.T.Baker®
Magnesium chloride hexahydrate
Formula: MgCl₂·6H₂O
Boiling Pt: 1412 °C (1013 hPa)
Melting Pt: 117 °C
Density: 1,57 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149781
CAS Number: 7791-18-6
EINECS: 232-094-6
Merck Index: 13,05684

Order Now

Specification Test Results

Meets ACS Specifications
Assay 99.0 - 102.0 %
Ammonium (NH4) max. 0.002 %
Barium (Ba) max. 0.005 %
Calcium (Ca) max. 0.01 %
Potassium (K) max. 0.005 %
Sodium (Na) max. 0.005 %
Strontium (Sr) max. 0.005 %
Insoluble Matter max. 0.005 %
Nitrate (NO3) max. 0.001 %
Sulfate (SO4) max. 0.002 %
Trace Impurities (in ppm)
Iron (Fe) max. 5
Manganese (Mn) max. 5
Heavy Metals (as Pb) max. 5
Phosphate (PO4) max. 5


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR