Lactic acid 85% 85%, BAKER ANALYZED® ACS, J.T.Baker®

Supplier: Avantor
Danger

Synonyms: DL-Lactic acid, 2-Hydroxypropionic acid, 2-Hydroxypropanoic acid

BAKR0194-01EA 113 SGD
BAKR0194-01 BAKR0194-03
Lactic acid 85% 85%, BAKER ANALYZED® ACS, J.T.Baker®
Lactic acid

The reagent generally available is a mixture of lactic acid (CH₃CHOHCOOH) and lactic acid lactate (C₆H₁₀O₅).

Formula: H₃CCH(OH)COOH
Boiling Pt: 122 °C (20 hPa)
Density: 1,20 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00004520
CAS Number: 50-21-5
EINECS: 200-018-0
UN: 3265
ADR: 8,II

Order Now

Specification Test Results

Meets ACS Reagent Chemical Requirements 85.0 - 90.0%
For Laboratory,Research,or Manufacturing Use ≤0.02%
ACS - Assay (as CH₃CHOHCOOH) 85.0 - 90.0 %
ACS - Residue after Ignition ≤ 0.02 %
ACS - Chloride (Cl) ≤ 0.001 %
ACS - Sulfate (SO₄) ≤ 0.002 %
ACS - Substances Darkened by H₂SO₄ Passes Test
Trace Impurities - ACS - Heavy Metals (as Pb) ≤ 5 ppm
Trace Impurities - Iron (Fe) ≤ 5 ppm
The reagent generally available is a mixture of lactic acid (CH₃CHOHCOOH) andlactic acid lactate (C₆H₁₀O₅).


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR