Sodium chloride, crystals, BAKER ANALYZED®, J.T.Baker®

Supplier: Avantor

BAKR0278.1000EA 40.9 SGD
BAKR0278.1000 BAKR0278.5000 BAKR0278.0500 BAKR0278.9050 BAKR0278.9025
Sodium chloride, crystals, BAKER ANALYZED®, J.T.Baker®
Sodium chloride
Formula: NaCl
Boiling Pt: 1413 °C (1013 hPa)
Melting Pt: 801 °C
Density: 2,16 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003477
CAS Number: 7647-14-5
EINECS: 231-598-3
Merck Index: 14,08599

Order Now

Specification Test Results

Assay min. 99.5 %
Barium (Ba) max. 0.002 %
Bromide (Br) max. 0.01 %
Calcium (Ca) max. 0.002 %
Iodide (I) max. 0.005 %
Potassium (K) max. 0.01 %
Magnesium (Mg) max. 0.001 %
Insoluble Matter max. 0.005 %
pH of 5% Solution at 25°C 5.0 - 8.0
Sulfate (SO4) max. 0.01 %
Trace Impurities (in ppm)
Arsenic (As) max. 3
Iron (Fe) max. 3
Heavy Metals (as Pb) max. 5
Phosphate (PO4) max. 5


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR