Sodium iodide, BAKER, Laboratory Reagent, J.T.Baker®

Supplier: Avantor
Warning

Synonyms: Hydriodic acid sodium salt, Sodium monoiodide, Ioduril

BAKR0294.0500EA 265 SGD
BAKR0294.0500
Sodium iodide, BAKER, Laboratory Reagent, J.T.Baker®
Sodium iodide
Formula: NaI
Boiling Pt: 1300 °C (1013 hPa)
Melting Pt: 662 °C
Density: 3,67 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003532
CAS Number: 7681-82-5
EINECS: 231-679-3
UN: 3077
ADR: 9,III
Merck Index: 13,08705

Order Now

Specification Test Results

Assay 99.0 - 100.5 %
Alkalinity passes test
Appearance of solution passes test
Iron (Fe) max. 20 ppm
Potassium (K) passes test
Heavy Metals (as Pb) max. 10 ppm
Identification passes test
Iodate (IO3) passes test
Limit of nitrate, nitrite and ammonia passes test
Loss on Drying max. 3.0 %
Organic Volatile Impurities passes test
Sulfate (SO4) max. 150 ppm
Thiosulfate and Barium passes test
Thiosulfates (as S2O3) passes test
Water (H2O) max. 2.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR