Acetic acid 80% solution, Biotech Reagent, J.T.Baker®

Supplier: Avantor
Danger

Synonyms: Vinegar acid, Methanecarboxylic acid, Ethanoic acid

BAKR0321-09EA 7880 SGD
BAKR0321-09
Acetic acid 80% solution, Biotech Reagent, J.T.Baker®
Acetic acid
Formula: H₃CCOOH
MW: 60.05 g/mol
Boiling Pt: 118
Melting Pt: 17
Flash Pt: 38.5
MDL Number: MFCD00036152
CAS Number: 64-19-7
UN: 2790
ADR: 8 II

Order Now

Specification Test Results

GMP Manufactured Product
CAUTION: For Manufacturing, processing or repackaging
Appearance Passes Test
Assay (CH₃COOH) 78.0 - 82.0 %
Chloride (Cl) ≤25 ppm
ACS - Heavy Metals (as Pb) ≤5 ppm
Iron (Fe) ≤5 ppm
Nonvolatile Residue (USP), mg/20 mL ≤1.0
Sulfate (SO₄) Passes Test
Made from U.S.P. Purified Water and Acetic Acid, Glacial, U.S.P. (Multi-Compendial) which meets BP, Ph.Eur. and JP Chemical Specifications
No Class 1,2,3 or other solvents are used or produced in the manufacturing or purification of the product.
Filtered through a 0.2 micron filter.

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR