Hydrochloric acid 6.0 N, Biotech Reagent, J.T.Baker®

Supplier: Avantor
Danger

Synonyms: Hydrochloric acid solution, Muriatic acid

Management of Change (MOC) category = R

BAKR0347-02EA 162 SGD
BAKR0347-02
Hydrochloric acid 6.0 N, Biotech Reagent, J.T.Baker®
Hydrochloric acid
Formula: HCl
MW: 36.46
Density: 1.06 g/cm³
MDL Number: MFCD00011324
CAS Number: 7647-01-0
UN: 1789
ADR: 8 II

Order Now

Specification Test Results

GMP Manufactured Product
CAUTION: For Manufacturing, processing or repackaging
Appearance (Clear, colorless liquid) Passes Test
Normality 5.95 - 6.05
Ammonium (NH₄) ≤ 3 ppm
Trace Impurities - Heavy Metals ≤ 0.5 ppm
Trace Impurities - Iron (Fe) ≤ 0.1 ppm
Sulfate (SO₄) ≤ 1 ppm
Made from U.S.P. Water for Injection (WFI) and Hydrochloric Acid, N.F.
(Multi-Compendial) which meets BP, Ph.Eur. and JP Chemical
Specifications
No Class 1,2,3 or other solvents are used or produced in the
manufacturing or purification of the product.
Filtered through a 0.2 micron filter.

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR