Sodium hydroxide 2.0 N in aqueous solution, Biotech Reagent, J.T.Baker®

Supplier: Avantor
Danger


Filtered through a 0.2 µm filter.

BAKR0390-07EA 569 SGD
BAKR0390-07
Sodium hydroxide 2.0 N in aqueous solution, Biotech Reagent, J.T.Baker®
Sodium hydroxide

Made from water for injection USP and sodium hydroxide pellets, NF (multi-compendial) which meets BP, Ph. Eur. and JP chemical specifications.

Formula: NaOH
Boiling Pt: 109 °C (1013 hPa)
Density: ≤1,11 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003548
CAS Number: 1310-73-2
EINECS: 215-185-5
UN: 1824
ADR: 8,II

Order Now

Specification Test Results

GMP Manufactured Product 1.9 - 2.1
Protect from air to avoid absorption of carbon dioxide. ≤5 ppm
CAUTION: For Manufacturing, processing or repackaging ≤1 ppm
Appearance (Clear, colorless liquid) Passes Test
Normality 1.9 - 2.1
Chloride (Cl) ≤ 5 ppm
ACS - Heavy Metals (as Pb) ≤ 1 ppm
Trace Impurities - Iron (Fe) ≤ 0.5 ppm
Endotoxin Concentration (EU/mL ≤ 2
Made from Water for Injection U.S.P. and Sodium Hydroxide pellets, N.F.(Multi-Compendial) which meets BP, Ph.Eur. and JP Chemical Specifications
No Class 1,2,3 or other solvents are used or produced in the manufacturing or purification of the product.
Filtered through a 0.2 micron filter.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR