Aluminium oxide, powder, BAKER ANALYZED® for chromatography, J.T.Baker®

Supplier: Avantor

Synonyms: Aluminium(III) oxide, Alumina C, Aluminum oxide

BAKR0537-01EA 156 SGD
BAKR0537-01
Aluminium oxide, powder, BAKER ANALYZED® for chromatography, J.T.Baker®
Aluminium oxide
Formula: Al₂O₃
Boiling Pt: 2980 °C (1013 hPa)
Melting Pt: 2045 °C
Density: 3,94 g/cm³ (20 °C)
MDL Number: MFCD00003424
CAS Number: 1344-28-1
EINECS: 215-691-6
REACH: 01-2119529248-35
Merck Index: 13,00355

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use 7.0 - 7.8
pH of 5% Slurry at 20°C 6.0 - 7.5
Adsorption, mg o-nitroaniline/g ≥0.05
Loss on Drying ≤5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR