Aluminium potassium sulphate dodecahydrate 98,0-102,0 EDTA titration, crystals, BAKER ANALYZED® ACS, J.T.Baker®

Supplier: Avantor

Synonyms: Aluminium Potassium sulphate, 12-Hydrate, Aluminum potassium sulphate dodecahydrate, Aluminium potassium bis(sulphate) dodecahydrate, Potassium alum, Alum potassium

BAKR0546-01EA 94.4 SGD
BAKR0546-01 BAKR0546-20 BAKR0546-05 BAKR0546-R
Aluminium potassium sulphate dodecahydrate 98,0-102,0 EDTA titration, crystals, BAKER ANALYZED® ACS, J.T.Baker®
Aluminium potassium sulphate dodecahydrate

Crystals. Lot analysis on label.

Formula: AlK(SO₄)₂·12H₂O
MW: 474.39 g/mol
Melting Pt: 92.5
Density: 1.72 g/cm³ (20 °C)
MDL Number: MFCD00149143
CAS Number: 7784-24-9
EINECS: 233-141-3
Merck Index: 12,00373

Order Now

Specification Test Results

Meets ACS Reagent Chemical Requirements 98.0 - 102.0%
For Laboratory,Research,or Manufacturing Use 99.0 - 100.5%
Assay (AlK(SO₄)₂·12H₂O) (by EDTA titrn) 98.0 - 102.0 %
Assay (dried basis) 99.0 - 100.5 %
Insoluble Matter ≤0.005 %
pH of 5% Solution at 25°C 2.0 - 4.0
Ammonium (NH₄) ≤0.005 %
Trace Impurities - Iron (Fe) ≤0.001 %
Sodium (Na) ≤0.02 %
Chloride (Cl) ≤5 ppm
Trace Impurities - ACS - Heavy Metals (as Pb) ≤5 ppm
Meets Reagent Specifications for testing USP/NF monographs

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR