Ammonium sulphate 99,0-100,5, granular ACS, FCC, J.T.Baker®

Supplier: Avantor
Warning

Synonyms: Sulphuric acid diammonium salt, Diammonium sulphate, Mascagnite

BAKR0795-08EA 6440 SGD
BAKR0795-08 BAKR0795-65 BAKR0795-12 BAKR0795-75
Ammonium sulphate 99,0-100,5, granular ACS, FCC, J.T.Baker®
Ammonium sulphate

  • Food GMP manufactured product

Formula: (NH₄)₂SO₄
MW: 132.14 g/mol
Melting Pt: 336…339 °C
Density: 1.78 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003391
CAS Number: 7783-20-2
EINECS: 231-984-1
REACH: 01-2119455044-46
Merck Index: 13,00559

Order Now

Specification Test Results

Meets ACS Reagent Chemical Requirements 99.0 - 100.5%
Food GMP Manufactured Product ≤0.005%
Meets F.C.C. Requirements ≤0.005%
CAUTION: For Manufacturing, processing or repackaging 5.0 - 6.0
Bulk Food Chemical ≤0.001%
ACS - Assay ((NH₄)₂SO₄) 99.0 - 100.5 %
ACS - Insoluble Matter ≤0.005 %
ACS - Residue on Ignition (ACS) ≤0.005 %
ACS - pH of 5% Solution at 25°C 5.0 - 6.0
ACS - Nitrate (NO₃) ≤0.001 %
Chloride (Cl) ≤3 ppm
Trace Impurities - Iron (Fe) ≤5 ppm
Phosphate (PO₄) ≤3 ppm
Heavy Metals (as Pb) ≤3 ppm
FCC - Assay ((NH₄)₂SO₄)(F.C.C.) 99.0 - 100.5 %
FCC - Identification A Passes Test
FCC - Identification B Passes Test
FCC - Residue on Ignition (FCC) ≤0.25 %
FCC - Lead (Pb) ≤3 mg/kg
FCC - Selenium (Se) ≤0.003 %

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR