Ammonium sulphate kit 99.0-100.5% NF endotoxin tested

Supplier: Avantor
Warning

Synonyms: Sulphuric acid diammonium salt, Diammonium sulphate, Mascagnite

BAKR0798-X1EA 14860 SGD
BAKR0798-X1
Ammonium sulphate kit 99.0-100.5% NF endotoxin tested
Ammonium sulphate
Formula: (NH₄)₂SO₄
MW: 132.14 g/mol
Melting Pt: 336…339 °C
Density: 1.77 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00003391
CAS Number: 7783-20-2
EINECS: 231-984-1
REACH: 01-2119455044-46
Merck Index: 13,00559

Order Now

Specification Test Results

GMP Manufactured Product
Meets N.F. Requirements
CAUTION: For Manufacturing, processing or repackaging
Bulk Pharmaceutical Chemical
Kit Components Meet Requirements Passes Test
This kit contains one each of Ammonium Sulfate product codes 0798-B1 and 0798-SA.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR