Ammonium sulphate, granules FCC, J.T.Baker®

Supplier: Avantor

Synonyms: Sulphuric acid diammonium salt, Diammonium sulphate, Mascagnite

BAKR0800-05EA 198 SGD
BAKR0800-05
Ammonium sulphate, granules FCC, J.T.Baker®
Ammonium sulphate
Formula: (NH₄)₂SO₄
MW: 132.14 g/mol
Melting Pt: 336…339 °C
Density: 1.78 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003391
CAS Number: 7783-20-2
EINECS: 231-984-1
REACH: 01-2119455044-46
Merck Index: 13,00559

Order Now

Specification Test Results

Food GMP Manufactured Product
Meets F.C.C. Requirements
CAUTION: For Manufacturing, processing or repackaging
Bulk Food Chemical 99.0 - 100.5 %
FCC - Assay 99.0 - 100.5 %
FCC - Identification Passes Test
FCC - Lead (Pb) ≤3 mg/kg
FCC - Residue on Ignition ≤0.25 %
FCC - Selenium (Se) ≤0.003 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR