Silver sulphate, BAKER ANALYZED®, Laboratory Reagent, J.T.Baker®

Supplier: Avantor
Danger

Synonyms: Disilver sulphate

BAKR1536.0025EA 247 SGD
BAKR1536.0025 BAKR1536.0250
Silver sulphate, BAKER ANALYZED®, Laboratory Reagent, J.T.Baker®
Silver sulphate
Formula: Ag₂SO₄
MW: 311.8 g/mol
Boiling Pt: 1085
Melting Pt: 655
Density: 5.45 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003407
CAS Number: 10294-26-5
EINECS: 233-653-7
Merck Index: 13,08603
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=dd7afaa8e9ae24bf040d40aed0c93763ce897080&vid=ae7b50634b53507047974c2157e5e38c2eb90a20&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay min. 99.5 %
Iron (Fe) max. 0.001 %
Insoluble Matter and Chloride max. 0.02 %
Nitrate (NO3) max. 0.01 %
Substances not Precipitated by HCl max. 0.03 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR