Chromium(III) chloride hexahydrate 99,0-105,0 Iodometry, BAKER ANALYZED®, J.T.Baker®

Supplier: Avantor
Warning

Synonyms: Chromium trichloride hexahydrate

BAKR1588-01EA 158 SGD
BAKR1588-01
Chromium(III) chloride hexahydrate 99,0-105,0 Iodometry, BAKER ANALYZED®, J.T.Baker®
Chromium (III) chloride hexahydrate
Formula: CrCl₃·6H₂O
MW: 266.45 g/mol
Melting Pt: 85
Density: 1.76 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149660
CAS Number: 10060-12-5
EINECS: 233-038-3
UN: 3260
ADR: 8 III
Merck Index: 13,02242

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use 99.0 - 105.0 %
Assay (CrCl₃6H₂O) (by iodometry) 99.0 - 105.0 %
Insoluble Matter ≤ 0.01 %
pH of 5% Solution at 25°C 2.0 - 3.0
Sulfate (SO₄) ≤ 0.01 %
Iron (Fe) ≤ 0.01 %

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR