Sodium dihydrogen phosphate dihydrate, BAKER, Laboratory Reagent, J.T.Baker®

Supplier: Avantor

Synonyms: Phosphoric acid (sodium hydrate), Sodium phosphate monobasic dihydrate, Sodium dihydrogen orthophosphate dihydrate

BAKR1768.1000EA 80.6 SGD
BAKR1768.1000
Sodium dihydrogen phosphate dihydrate, BAKER, Laboratory Reagent, J.T.Baker®
Sodium dihydrogen phosphate dihydrate
Formula: NaH₂PO₄·2H₂O
Boiling Pt: ∼650 °C (1013 hPa)
Melting Pt: 60 °C
Density: 1,91 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149209
CAS Number: 13472-35-0
EINECS: 231-449-2
REACH: 01-2119489796-13
Merck Index: 12,08806

Order Now

Specification Test Results

Assay min. 98 %
Trace Impurities (in ppm)
Arsenic (As) max. 2


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR