1-Heptanesulphonic acid sodium salt ≥98.0% (by acidimetry) for HPLC, for ion pair chromatography, J.T.Baker®

Supplier: Avantor
Warning

Synonyms: Sodium heptane-1-sulphonate, Sodium 1-heptanesulphonate

BAKR2173-05EA 139 SGD
BAKR2173-05
1-Heptanesulphonic acid sodium salt ≥98.0% (by acidimetry) for HPLC, for ion pair chromatography, J.T.Baker®
1-Heptanesulphonic acid sodium salt

For ion-pair chromatography of basic compounds. UV cutoff 210 nm.

Formula: H₃C(CH₂)₅CH₂SO₃Na
MW: 202.25 g/mol
Melting Pt: 300
Storage Temperature: Ambient
MDL Number: MFCD00007543
CAS Number: 22767-50-6
EINECS: 245-210-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=95c09a5691140bab7446d88919f01593e8f0b9b2&vid=7c237cde5db01d0567ff19964ca3287434747480&prid=web&clid=SAPDAM

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use ≥98.0%
Assay (as C₇H₁₅SO₃Na) (by Acidimetry) ≥98.0%
UV Absorbance of 0.25 M Solution - 250 nm ≤0.050
UV Absorbance of 0.25 M Solution - 240 nm ≤0.050
UV Absorbance of 0.25 M Solution - 230 nm ≤0.050
UV Absorbance of 0.25 M Solution - 220 nm ≤0.060
UV Absorbance of 0.25 M Solution - 210 nm ≤0.080
UV Absorbance of 0.25 M Solution - 200 nm ≤0.200


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR