Lithium chloride 99,0 Argentometric titration, granular, BAKER ANALYZED® ACS, J.T.Baker®

Supplier: Avantor
Warning

BAKR2370-01EA 268 SGD
BAKR2370-01 BAKR2370-07 BAKR2370-05
Lithium chloride 99,0 Argentometric titration, granular, BAKER ANALYZED® ACS, J.T.Baker®
Lithium chloride
Formula: LiCl
MW: 42.39 g/mol
Boiling Pt: 1382
Melting Pt: 614
Storage Temperature: Ambient
MDL Number: MFCD00011078
CAS Number: 7447-41-8
Merck Index: 13,05550

Order Now

Specification Test Results

Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use = 99.0 %
ACS - Assay (LiCl) (by Ag titrn) ≥ 99.0 %
ACS - Insoluble Matter ≤ 0.01 %
ACS - Loss on Drying ≤ 1.0 %
ACS - Titrable Base (meq/g) ≤ 0.008
ACS - Sulfate (SO₄) ≤ 0.01 %
ACS - Barium (Ba) ≤ 0.002 %
ACS - Heavy Metals (as Pb) ≤ 0.002 %
ACS - Iron (Fe) ≤ 0.001 %
ACS - Calcium (Ca) ≤ 0.01 %
ACS - Potassium (K) ≤ 0.01 %
ACS - Sodium (Na) ≤ 0.20 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR