Magnesium sulphate heptahydrate 99.0-100.5% (by anhydrous basis), crystals USP, Multi-Compendial, J.T.Baker®

Supplier: Avantor

Synonyms: sulphuric acid magnesium salt heptahydrate, Epsom salt, Bitter salt

BAKR2504-01EA 117 SGD
BAKR2504-01 BAKR2504-05 BAKR2504-07 BAKR2504-08
Magnesium sulphate heptahydrate 99.0-100.5% (by anhydrous basis), crystals USP, Multi-Compendial, J.T.Baker®
Magnesium sulphate heptahydrate

  • GMP manufactured product

Formula: MgSO₄·7H₂O
MW: 246.48 g/mol
Melting Pt: 1124
Density: 1.67 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00149785
CAS Number: 10034-99-8
EINECS: 231-298-2
Merck Index: 13,05715

Order Now

Specification Test Results

GMP Manufactured Product Passes Test
Meets B.P. Chemical Specifications 5.0 - 9.2
Meets E.P. Chemical Specifications 40.0 - 52.0%
Meets U.S.P Requirements ≤0.014%
Preserve in well-closed containers. ≤0.5 ppm
CAUTION: For Manufacturing, processing or repackaging ≤0.003%
Bulk Pharmaceutical Chemical 99.0 - 100.5%
USP - Identification Passes Test
USP - pH (1 in 20) 5.0 - 9.2
USP - Loss on Ignition 40.0 - 52.0 %
USP - Chloride (Cl) ≤ 0.014 %
USP - Iron (Fe) ≤ 0.5 ppm
USP - Selenium (Se) ≤ 0.003 %
USP - Assay (MgSO₄) (anhydrous basis) 99.0 - 100.5 %
USP - Endotoxin Concentration (EU/g) ≤ 2.5
EP/BP - Identification A Passes Test
EP/BP - Identification B Passes Test
EP/BP - Appearance of Solution Passes Test
EP/BP - Acidity or Alkalinity Passes Test
EP/BP - Chloride (Cl) ≤ 300 ppm
EP/BP - Arsenic (As) ≤ 2 ppm
EP/BP - Iron (Fe) ≤ 20 ppm
EP/BP - Loss on Drying 48.0 - 52.0 %
EP/BP - Assay (MgSO₄) (anhydrous basis) 99.0 - 100.5 %
Solution Test Passes Test
No Class 1,2,3 or other solvents are used or produced in the
manufacturing or purification of the product.
Metallic Residues: No metal catalysts or metal reagents, as defined by
EMA Guideline EMEA/CHMP/SWP/4446/2000 , are used in the production of
this material.
This product is intended for use in preparing parenteral and
nonparenteral dosage forms.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR