1-Octanesulphonic acid sodium salt ≥98.0% for ion pair chromatography, for ion pair chromatography, J.T.Baker®

Supplier: Avantor
Warning

Synonyms: Sodium octane-1-sulphonate

BAKR2818-05EA 169 SGD
BAKR2818-05
1-Octanesulphonic acid sodium salt ≥98.0% for ion pair chromatography, for ion pair chromatography, J.T.Baker®
1-Octanesulphonic acid sodium salt
Formula: C₈H₁₇NaO₃S
MW: 216.28 g/mol
Melting Pt: > 300 °C
Storage Temperature: Ambient
MDL Number: MFCD00007544
CAS Number: 5324-84-5
EINECS: 226-195-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1244cb11db07775dec2e193823e40080024b7036&vid=32d95cd9e321818e2f94c8261c4b4e6180ee0fc7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use = 98.0 %
Assay (as C₈H₁₇SO₃Na) (by acidimetry) ≥98.0 %
UV Absorbance of 0.25 M Solution - 250 nm ≤0.050
UV Absorbance of 0.25 M Solution - 240 nm ≤0.050
UV Absorbance of 0.25 M Solution - 230 nm ≤0.050
UV Absorbance of 0.25 M Solution - 220 nm ≤0.060
UV Absorbance of 0.25 M Solution - 210 nm ≤0.080
UV Absorbance of 0.25 M Solution - 200 nm ≤0.200
Meets Reagent Specifications for testing USP/NF monographs


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR