1-Pentanesulphonic acid sodium salt ≥98% for ion pair chromatography, J.T.Baker®

Supplier: Avantor

BAKR2841-05EA 179 SGD
BAKR2841-05 BAKR2841-06
1-Pentanesulphonic acid sodium salt ≥98% for ion pair chromatography, J.T.Baker®
1-Pentanesulphonic acid sodium salt
Formula: C₅H₁₁O₃SNa
Melting Pt: > 300 °C
Storage Temperature: Ambient
MDL Number: MFCD00007541
CAS Number: 22767-49-3
EINECS: 245-208-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ddb1c23c8a6fe3c156a3aacb62721e1d1a34efb0&vid=1936dcdb47ba374ce21fd758660f7ca6ba95d254&prid=web&clid=SAPDAM

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use = 98.0 %
Assay (as C₅H₁₁SO₃Na) (by acidimetry) ≥ 98.0 %
UV Absorbance of 0.25 M Solution - 250 nm ≤ 0.050
UV Absorbance of 0.25 M Solution - 240 nm ≤ 0.050
UV Absorbance of 0.25 M Solution - 230 nm ≤ 0.050
UV Absorbance of 0.25 M Solution - 220 nm ≤ 0.060
UV Absorbance of 0.25 M Solution - 210 nm ≤ 0.080
UV Absorbance of 0.25 M Solution - 200 nm ≤ 0.200


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR